CAS RN 723262-95-1
IUPAC Name:
6,6-dimethyl-5-phenyl-5,6-dihydro-2H-1,4-oxazin-2-one
Synonyms:
6,6-dimethyl-5-phenyl-5H-1,4-oxazin-2-one
Molecular Weight (g/mol):
203.24
Molecular Formula:
C12H13NO2
InChi Key:
AISCPSRYGCFAQP-UHFFFAOYNA-N
SMILES:
CC1(C)OC(=O)C=NC1C1=CC=CC=C1
1
–
1
of
1
results
1
(5S)-5,6-Dihydro-6,6-dimethyl-5-phenyl-2H-1,4-oxazin-2-one 95.0+%, TCI America™
CAS: 723262-95-1 Molecular Formula: C12H13NO2 Molecular Weight (g/mol): 203.241 InChI Key: AISCPSRYGCFAQP-NSHDSACASA-N PubChem CID: 11367698 IUPAC Name: (3S)-2,2-dimethyl-3-phenyl-3H-1,4-oxazin-6-one SMILES: CC1(C(N=CC(=O)O1)C2=CC=CC=C2)C