accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 723284-79-5

IUPAC Name: 3-amino-3-(3-fluorophenyl)propanoic acid
Synonyms: 3-amino-3-(3-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 183.18
Molecular Formula: C9H10FNO2
InChi Key: GZNJUJNKZBHINS-UHFFFAOYNA-N
SMILES: NC(CC(O)=O)C1=CC(F)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)