accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7236-47-7

IUPAC Name: (3E)-3-methyl-4-phenylbut-3-enamide
Synonyms: β-benzalbutyramide
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: KAJZGRFYZKWYDX-VQHVLOKHSA-N
SMILES: C\C(CC(N)=O)=C/C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results