accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH3

CAS RN 72388-18-2

IUPAC Name: 2-dodecylhexadecan-1-ol
Synonyms: 2-dodecylhexadecan-1-ol
Molecular Weight (g/mol): 410.77
Molecular Formula: C28H58O
InChi Key: DEMBLPGWNXUBIQ-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCC(CO)CCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results