accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7240-38-2

IUPAC Name: sodium (2S,5R,6R)-3,3-dimethyl-6-(5-methyl-3-phenyl-1,2-oxazole-4-amido)-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate
Synonyms: oxacillin sodium monohydrate oxacillin sodium salt monohydrate
Molecular Weight (g/mol): 441.43
Molecular Formula: C19H20N3NaO6S
InChi Key: ZVIYWUUZWWBNMB-VICXVTCVSA-M
SMILES: O.[Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)
Percent Purity 
  • (2)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)