accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOO

CAS RN 72418-40-7

IUPAC Name: 3-phenyl-1,2-oxazole-5-carbaldehyde
Synonyms: 3-phenyl-1,2-oxazole-5-carbaldehyde
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: SZXMDUKWIYIKKS-UHFFFAOYSA-N
SMILES: O=CC1=CC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results