accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72432-03-2

IUPAC Name: (2R,3R,4R,5S)-1-(2-hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol
Synonyms: miglitol miglitol (substance) diastabol miglitol preparation miglitolum glyset plumarol lacer brand of miglitol miglitol (product)
Molecular Weight (g/mol): 207.23
Molecular Formula: C8H17NO5
InChi Key: IBAQFPQHRJAVAV-ULAWRXDQSA-N
SMILES: OCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results