accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOO

CAS RN 7245-02-5

IUPAC Name: 3-methoxy-2-phenyl-4H-chromen-4-one
Synonyms: 3-methoxy-2-phenylchromen-4-one
Molecular Weight (g/mol): 252.27
Molecular Formula: C16H12O3
InChi Key: ZAIANDVQAMEDFL-UHFFFAOYSA-N
SMILES: COC1=C(OC2=CC=CC=C2C1=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)