accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7245-21-8

IUPAC Name: λ²-iron(2+) dimethanolate
Synonyms: λ²-iron(2+) bis(methoxide)
Molecular Weight (g/mol): 117.91
Molecular Formula: C2H6FeO2
InChi Key: VAPRHKOWFRYFTF-UHFFFAOYSA-N
SMILES: [Fe++].C[O-].C[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results