CAS RN 72459-47-3
IUPAC Name:
1-[(3-bromophenyl)methyl]-1H-imidazole
Synonyms:
1-[(3-bromophenyl)methyl]imidazole
Molecular Weight (g/mol):
237.1
Molecular Formula:
C10H9BrN2
InChi Key:
BGYZLOIBJCHCTI-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(CN2C=CN=C2)=C1
1
–
1
of
1
results
1
1-(3-Bromobenzyl)-1H-imidazole, ≥97%, Thermo Scientific™
CAS: 72459-47-3 Molecular Formula: C10H9BrN2 Molecular Weight (g/mol): 237.10 MDL Number: MFCD08271919 InChI Key: BGYZLOIBJCHCTI-UHFFFAOYSA-N Synonym: 1-3-bromobenzyl-1h-imidazole,1-3-bromophenyl methyl imidazole,1-3-bromobenzyl imidazole,1-3-bromophenyl methyl-1h-imidazole,1h-imidazole,1-3-bromophenyl methyl,d07jmo,3-bromophenyl methyl imidazole PubChem CID: 10220337 IUPAC Name: 1-[(3-bromophenyl)methyl]imidazole SMILES: BrC1=CC=CC(CN2C=CN=C2)=C1