accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClClN

CAS RN 7250-67-1

IUPAC Name: hydrogen 1-(2-chloroethyl)pyrrolidine chloride
Synonyms: hydrogen 1-(2-chloroethyl)pyrrolidine chloride
Molecular Weight (g/mol): 170.08
Molecular Formula: C6H13Cl2N
InChi Key: FSNGFFWICFYWQC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClCCN1CCCC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (4)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)