accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3COO(S)O(R)O(S)O(S)HO(S)HO(R)OH(S)HO(S)OH(R)OH

CAS RN 72520-92-4

IUPAC Name: 1-(4-methoxy-2-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2S,3R,4S,5S)-3,4,5-trihydroxyoxan-2-yl]oxy}methyl)oxan-2-yl]oxy}phenyl)ethan-1-one
Synonyms: paeonolide
Molecular Weight (g/mol): 460.43
Molecular Formula: C20H28O12
InChi Key: IDZZECHGWAZTIB-NYBIBFQCSA-N
SMILES: COC1=CC=C(C(C)=O)C(O[C@@H]2O[C@H](CO[C@@H]3OC[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)