accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 725211-89-2

IUPAC Name: 1-{1-[(furan-2-yl)methyl]piperidin-4-yl}methanamine
Synonyms: 1-[1-(furan-2-ylmethyl)piperidin-4-yl]methanamine
Molecular Weight (g/mol): 194.28
Molecular Formula: C11H18N2O
InChi Key: GKCIOGOJFDDOJI-UHFFFAOYSA-N
SMILES: NCC1CCN(CC2=CC=CO2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results