accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONN

CAS RN 7254-34-4

IUPAC Name: 2-methoxypyridine-3-carbonitrile
Synonyms: 2-methoxypyridine-3-carbonitrile
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: IHWZCGMZILLKFD-UHFFFAOYSA-N
SMILES: COC1=NC=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results