accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOOH

CAS RN 72580-53-1

IUPAC Name: pyrrolidine-3-carboxylic acid
Synonyms: pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: JAEIBKXSIXOLOL-UHFFFAOYNA-N
SMILES: OC(=O)C1CCNC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)