accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2ClFFF

CAS RN 72587-18-9

IUPAC Name: 2-chloro-5-(trifluoromethyl)pyridin-3-amine
Synonyms: 2-chloro-5-(trifluoromethyl)pyridin-3-amine
Molecular Weight (g/mol): 196.56
Molecular Formula: C6H4ClF3N2
InChi Key: HABKYPVMMHBOIW-UHFFFAOYSA-N
SMILES: NC1=C(Cl)N=CC(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results