CAS RN 72599-27-0
IUPAC Name:
(2R,3R,4R,5S)-1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol
Synonyms:
miglustat butyldeoxynojirimycin n-butyl dnj zavesca BuDNJ N-butyl-1-deoxynojirimycin miglustatum N-butylmoranoline NB-dnj n-butyldeoxynojirimycin
Molecular Weight (g/mol):
219.28
Molecular Formula:
C10H21NO4
InChi Key:
UQRORFVVSGFNRO-UTINFBMNSA-N
SMILES:
CCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO
1
–
1
of
1
results
1
N-Butyldeoxynojirimycin, 98%, Thermo Scientific Chemicals
CAS: 72599-27-0 Molecular Formula: C10H21NO4 Molecular Weight (g/mol): 219.28 MDL Number: MFCD00272581 InChI Key: UQRORFVVSGFNRO-UTINFBMNSA-N PubChem CID: 51634 ChEBI: CHEBI:50381 IUPAC Name: (2R,3R,4R,5S)-1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol SMILES: CCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO