CAS RN 72658-10-7
CAS RN 72658-10-7
IUPAC Name:
{[1-(tert-butoxy)prop-1-en-1-yl]oxy}trimethylsilane
Synonyms:
{[1-(tert-butoxy)prop-1-en-1-yl]oxy}trimethylsilane
Molecular Weight (g/mol):
202.37
Molecular Formula:
C10H22O2Si
InChi Key:
OJYHIAHTMZIVKY-UHFFFAOYSA-N
SMILES:
CC=C(OC(C)(C)C)O[Si](C)(C)C
1
–
1
of
1
results
1
(1E)-1-tert-Butoxy-1-(trimethylsilyloxy)propene 92.0+%, TCI America™

CAS: 72658-10-7 Molecular Formula: C10H22O2Si Molecular Weight (g/mol): 202.369 InChI Key: OJYHIAHTMZIVKY-UHFFFAOYSA-N PubChem CID: 57353179 IUPAC Name: trimethyl-[1-[(2-methylpropan-2-yl)oxy]prop-1-enoxy]silane SMILES: CC=C(OC(C)(C)C)O[Si](C)(C)C