accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72678-19-4

IUPAC Name: 2,6-dibromo-4-(trifluoromethyl)aniline
Synonyms: 2,6-dibromo-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 318.92
Molecular Formula: C7H4Br2F3N
InChi Key: DRSMEHXBOXHXDX-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(C=C1Br)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results