accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72692-99-0

IUPAC Name: 3-(1H-pyrrol-1-yl)pyridine
Synonyms: 3-(pyrrol-1-yl)pyridine
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: SPFWVEKHAJYTGG-UHFFFAOYSA-N
SMILES: C1=CN(C=C1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results