accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)N(R)O(S)(R)NNOHNH2HOOPOPHOOOOOHK

CAS RN 72696-48-1

IUPAC Name: potassium (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-({[(hydrogen phosphonatooxy)(hydroxy)phosphoryl]oxy}methyl)oxolane-3,4-diol
Synonyms: potassium (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-({[hydrogen phosphonatooxy(hydroxy)phosphoryl]oxy}methyl)oxolane-3,4-diol
Molecular Weight (g/mol): 465.29
Molecular Formula: C10H14KN5O10P2
InChi Key: ZNCWUOPIJTUALR-GWKNMROSNA-M
SMILES: [K+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)