CAS RN 727-71-9
IUPAC Name:
2-phenyl-1-(2,4,6-trihydroxyphenyl)ethan-1-one
Synonyms:
2-phenyl-1-(2,4,6-trihydroxyphenyl)ethanone
Molecular Weight (g/mol):
244.25
Molecular Formula:
C14H12O4
InChi Key:
SLHBRIIHMDJIBT-UHFFFAOYSA-N
SMILES:
OC1=CC(O)=C(C(=O)CC2=CC=CC=C2)C(O)=C1
1
–
1
of
1
results
1
2',4',6'-Trihydroxy-2-phenylacetophenone 96.0+%, TCI America™
CAS: 727-71-9 Molecular Formula: C14H12O4 Molecular Weight (g/mol): 244.25 MDL Number: MFCD00205539 InChI Key: SLHBRIIHMDJIBT-UHFFFAOYSA-N Synonym: Benzyl 2,4,6-Trihydroxyphenyl Ketone PubChem CID: 689111 IUPAC Name: 2-phenyl-1-(2,4,6-trihydroxyphenyl)ethan-1-one SMILES: OC1=CC(O)=C(C(=O)CC2=CC=CC=C2)C(O)=C1