accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7270-63-5

IUPAC Name: 2,2-dimethyl-5-(λ⁵-diazynylidene)-1,3-dioxane-4,6-dione
Synonyms: 2,2-dimethyl-5-(λ⁵-diazynylidene)-1,3-dioxane-4,6-dione
Molecular Weight (g/mol): 170.12
Molecular Formula: C6H6N2O4
InChi Key: YRCVEMYVKSXPLM-UHFFFAOYSA-N
SMILES: CC1(C)OC(=O)C(=[N+]=[N-])C(=O)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results