accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72707-66-5

IUPAC Name: 2-(bromomethyl)prop-2-enoic acid
Synonyms: 2-(bromomethyl)prop-2-enoic acid
Molecular Weight (g/mol): 164.99
Molecular Formula: C4H5BrO2
InChi Key: NOOYFQLPKUQDNE-UHFFFAOYSA-N
SMILES: OC(=O)C(=C)CBr
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)