accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72752-52-4

IUPAC Name: 2-(piperidin-1-yl)benzonitrile
Synonyms: 2-(piperidin-1-yl)benzonitrile
Molecular Weight (g/mol): 186.26
Molecular Formula: C12H14N2
InChi Key: MEBVSLLKZSAIGK-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1N1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)