accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH2SH2NN

CAS RN 72792-54-2

IUPAC Name: 1,3-thiazole-2,4-diamine hydrochloridyl
Synonyms: 1,3-thiazole-2,4-diamine hydrochloridyl
Molecular Weight (g/mol): 150.6
Molecular Formula: C3H5ClN3S
InChi Key: QCFHKEYJZBLZKK-UHFFFAOYSA-N
SMILES: [Cl].NC1=NC(N)=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)