accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72798-32-4

IUPAC Name: 6,8-dichloro-4-oxo-4H-chromene-3-carbonitrile
Synonyms: 6,8-dichloro-4-oxochromene-3-carbonitrile
Molecular Weight (g/mol): 240.04
Molecular Formula: C10H3Cl2NO2
InChi Key: YERQUFAYRCSXEQ-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C2OC=C(C#N)C(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results