accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 728039-63-2

IUPAC Name: N,N-bis({[1,1'-biphenyl]-4-yl})-4'-bromo-[1,1'-biphenyl]-4-amine
Synonyms: N,N-bis({[1,1'-biphenyl]-4-yl})-4'-bromo-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 552.52
Molecular Formula: C36H26BrN
InChi Key: VBQIVFTUXFKGKW-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results