CAS RN 72804-96-7
CAS RN 72804-96-7
IUPAC Name:
amino diphenylphosphinate
Synonyms:
amino diphenylphosphinate
Molecular Weight (g/mol):
233.21
Molecular Formula:
C12H12NO2P
InChi Key:
SJECIYLGISUNRO-UHFFFAOYSA-N
SMILES:
NOP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
O-(Diphenylphosphinyl)hydroxylamine, 96%
CAS: 72804-96-7 Molecular Formula: C12H12NO2P Molecular Weight (g/mol): 233.21 InChI Key: SJECIYLGISUNRO-UHFFFAOYSA-N Synonym: o-diphenylphosphinylhydroxylamine,aminooxy diphenylphosphine oxide,o-diphenylphosphinyl hydroxylamine,o-diphenylphosphoryl hydroxylamine,amino diphenylphosphinate,hydroxylamine, o-diphenylphosphinyl,unii-2o9pf20t75,aminooxy diphenyl phosphine oxide,o-diphenylphosphinyl hydroxylamine mi PubChem CID: 10955453 IUPAC Name: O-diphenylphosphorylhydroxylamine SMILES: NOP(=O)(C1=CC=CC=C1)C1=CC=CC=C1