accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNSOOH2N

CAS RN 72811-73-5

IUPAC Name: 4-[(3-methylphenyl)amino]pyridine-3-sulfonamide
Synonyms: 4-[(3-methylphenyl)amino]pyridine-3-sulfonamide
Molecular Weight (g/mol): 263.32
Molecular Formula: C12H13N3O2S
InChi Key: ZXPCUGWAKUIOOF-UHFFFAOYSA-N
SMILES: CC1=CC(NC2=C(C=NC=C2)S(N)(=O)=O)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results