accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOOOCH3CH2

CAS RN 72829-09-5

IUPAC Name: 12-[(2-methylprop-2-enoyl)oxy]dodecyl 2-methylprop-2-enoate
Synonyms: 12-[(2-methylprop-2-enoyl)oxy]dodecyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 338.49
Molecular Formula: C20H34O4
InChi Key: HYQASEVIBPSPMK-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCCCCCCCCCCCOC(=O)C(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results