accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7283-69-4

IUPAC Name: 1,4-bis(2-methylpropyl) (2E)-but-2-enedioate
Synonyms: fumaric acid, diisobutyl ester
Molecular Weight (g/mol): 228.29
Molecular Formula: C12H20O4
InChi Key: RSRICHZMFPHXLE-AATRIKPKSA-N
SMILES: CC(C)COC(=O)\C=C\C(=O)OCC(C)C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results