accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOOOCH3H3C(E)

CAS RN 7283-70-7

IUPAC Name: 1,4-bis(propan-2-yl) (2E)-but-2-enedioate
Synonyms: 1,4-diisopropyl (2E)-but-2-enedioate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: FNMTVMWFISHPEV-AATRIKPKSA-N
SMILES: CC(C)OC(=O)\C=C\C(=O)OC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results