accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72835-25-7

IUPAC Name: 5-methylthiophene-2-carbonitrile
Synonyms: 5-methylthiophene-2-carbonitrile
Molecular Weight (g/mol): 123.17
Molecular Formula: C6H5NS
InChi Key: RBQRZWYCXAXPIN-UHFFFAOYSA-N
SMILES: CC1=CC=C(S1)C#N
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results