accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOClH2NCl

CAS RN 7286-84-2

IUPAC Name: methyl 3-amino-2,5-dichlorobenzoate
Synonyms: chloramben, methyl ester
Molecular Weight (g/mol): 220.05
Molecular Formula: C8H7Cl2NO2
InChi Key: DTSSCQVCVYZGSI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC(N)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results