accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN

CAS RN 72928-54-2

IUPAC Name: 4-(4-ethylcyclohexyl)benzonitrile
Synonyms: 4-(4-ethylcyclohexyl)benzonitrile
Molecular Weight (g/mol): 213.32
Molecular Formula: C15H19N
InChi Key: BBHJTCADCKZYSO-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results