accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72934-37-3

IUPAC Name: 1-(4-chlorophenyl)cyclopropane-1-carboxylic acid
Synonyms: 1-(4-chlorophenyl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 196.63
Molecular Formula: C10H9ClO2
InChi Key: YAHLWSGIQJATGG-UHFFFAOYSA-N
SMILES: OC(=O)C1(CC1)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)