accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7295-44-5

IUPAC Name: 1-(4-bromophenyl)pentan-1-one
Synonyms: 1-(4-bromophenyl)pentan-1-one
Molecular Weight (g/mol): 241.13
Molecular Formula: C11H13BrO
InChi Key: STYJKBMRWQQJIS-UHFFFAOYSA-N
SMILES: CCCCC(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results