accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOOH3C

CAS RN 72955-82-9

IUPAC Name: [2-(2-methoxyphenoxy)ethyl](methyl)amine
Synonyms: [2-(2-methoxyphenoxy)ethyl](methyl)amine
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: VOIPEKOXTFLQNY-UHFFFAOYSA-N
SMILES: CNCCOC1=CC=CC=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results