accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2N(S)NHO(S)HNNNHOH2NOHO

CAS RN 72957-37-0

IUPAC Name: (2S)-6-amino-2-[(2S)-2-(2-aminoacetamido)-3-(1H-imidazol-5-yl)propanamido]hexanoic acid
Synonyms: copper peptide ghk-cu
Molecular Weight (g/mol): 340.38
Molecular Formula: C14H24N6O4
InChi Key: MVORZMQFXBLMHM-QWRGUYRKSA-N
SMILES: NCCCC[C@H](NC(=O)[C@H](CC1=CN=CN1)NC(=O)CN)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)