CAS RN 7299-91-4
IUPAC Name:
butyl (2E)-but-2-enoate
Synonyms:
butyl (2E)-but-2-enoate
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
GWCTUKHUBWMAMI-GQCTYLIASA-N
SMILES:
CCCCOC(=O)\C=C\C
1
–
1
of
1
results
1
Butyl Crotonate 97.0+%, TCI America™
CAS: 7299-91-4 Molecular Formula: C8H14O2 Molecular Weight (g/mol): 142.20 MDL Number: MFCD00053794 InChI Key: GWCTUKHUBWMAMI-GQCTYLIASA-N Synonym: Crotonic Acid Butyl Ester PubChem CID: 5366039 IUPAC Name: butyl (2E)-but-2-enoate SMILES: CCCCOC(=O)\C=C\C