CAS RN 73-32-5
L-Isoleucine (White Crystals or Crystalline Powder), Fisher BioReagents™
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
Thermo Scientific Chemicals L-Isoleucine, 99%
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
Thermo Scientific Chemicals L-Isoleucine, 99%
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
L-Isoleucine, 99%, MP Biomedicals
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
Thermo Scientific Chemicals L-Isoleucine, Cell Culture Reagent
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
L-Isoleucine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
L-Isoleucine is an isomer of L-leucine and an essential amino acid. It is synthesized from threonine and is a branched-chain hydrophobic amino acid.
L-Isoleucine 98.0+%, TCI America™
CAS: 73-32-5 Molecular Formula: C6H13NO2 Molecular Weight (g/mol): 131.18 MDL Number: MFCD00064222 MFCD00004268 InChI Key: AGPKZVBTJJNPAG-WHFBIAKZSA-N Synonym: l-isoleucine,isoleucine,2s,3s-2-amino-3-methylpentanoic acid,s-isoleucine,s,s-isoleucine,2s,3s-isoleucine,2-amino-3-methylvaleric acid,erythro-l-isoleucine,l-+-isoleucine,l-ile PubChem CID: 6306 ChEBI: CHEBI:17191 IUPAC Name: (2S,3S)-2-amino-3-methylpentanoic acid SMILES: CC[C@H](C)[C@H](N)C(O)=O
Medisca Isoleucine (L), USP
Isoleucine (L), USP comes as white, practically odorless crystals. It is soluble in water and slightly soluble in hot alcohol.
Thermo Scientific Production Chemicals and Services L-Isoleucine USP/EP/JP/FCC, Ajinomoto
CAS: 73-32-5 Molecular Weight (g/mol): 131.17