accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOClNOO

CAS RN 73033-58-6

IUPAC Name: (5-chloro-2-nitrophenyl)methanol
Synonyms: (5-chloro-2-nitrophenyl)methanol
Molecular Weight (g/mol): 187.58
Molecular Formula: C7H6ClNO3
InChi Key: ULYZTHQGJXPEFT-UHFFFAOYSA-N
SMILES: OCC1=CC(Cl)=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results