accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7305-71-7

IUPAC Name: 5-methyl-1,3-thiazol-2-amine
Synonyms: 5-methyl-1,3-thiazol-2-amine
Molecular Weight (g/mol): 114.17
Molecular Formula: C4H6N2S
InChi Key: GUABFMPMKJGSBQ-UHFFFAOYSA-N
SMILES: CC1=CN=C(N)S1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)