accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7311-34-4

IUPAC Name: 3,5-dimethoxybenzaldehyde
Synonyms: 3,5-dimethoxy-benzaldehyde
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: VFZRZRDOXPRTSC-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC(OC)=C1
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (7)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)