CAS RN 7311-70-8
IUPAC Name:
2,5-dibromothiophene-3-carboxylic acid
Synonyms:
2,5-dibromothiophene-3-carboxylic acid
Molecular Weight (g/mol):
285.94
Molecular Formula:
C5H2Br2O2S
InChi Key:
PZBUYFPAASJYSI-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(Br)SC(Br)=C1
1
–
1
of
1
results
1
2,5-Dibromothiophene-3-carboxylic Acid 96.0+%, TCI America™
CAS: 7311-70-8 Molecular Formula: C5H2Br2O2S Molecular Weight (g/mol): 285.94 MDL Number: MFCD08445656 InChI Key: PZBUYFPAASJYSI-UHFFFAOYSA-N Synonym: 2,5-Dibromo-3-thenoic Acid PubChem CID: 268907 IUPAC Name: 2,5-dibromothiophene-3-carboxylic acid SMILES: OC(=O)C1=C(Br)SC(Br)=C1