accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSBr

CAS RN 7312-10-9

IUPAC Name: 5-bromo-1-benzothiophene-2-carboxylate
Synonyms: 5-bromo-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 256.09
Molecular Formula: C9H4BrO2S
InChi Key: ONNFNEFYXIPHCA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=CC(Br)=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results