accessibility menu, dialog, popup

UserName

Viewing profile as user:
Mn

CAS RN 73138-26-8

IUPAC Name: Bis(cyclopentadienyl)manganese
Synonyms: —
Molecular Weight (g/mol): 185.13
Molecular Formula: C10H10Mn
InChi Key: —
SMILES: [Mn](c1cccc1)c1cccc1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results