accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7314-32-1

IUPAC Name: (2S)-2-amino-4-methanesulfonylbutanoic acid
Synonyms: methionine sulfone
Molecular Weight (g/mol): 181.21
Molecular Formula: C5H11NO4S
InChi Key: UCUNFLYVYCGDHP-BYPYZUCNSA-N
SMILES: CS(=O)(=O)CC[C@H](N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)