accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOOClNNOSCl

CAS RN 73151-29-8

IUPAC Name: 1-[2-(2,4-dichlorophenyl)-2-{[4-(phenylsulfanyl)phenyl]methoxy}ethyl]-1H-imidazole; nitric acid
Synonyms: fenticonazole
Molecular Weight (g/mol): 518.41
Molecular Formula: C24H21Cl2N3O4S
InChi Key: FJNRUWDGCVDXLU-UHFFFAOYNA-N
SMILES: O[N+]([O-])=O.ClC1=CC=C(C(CN2C=CN=C2)OCC2=CC=C(SC3=CC=CC=C3)C=C2)C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results